missing translation for 'onlineSavingsMsg'
Learn More

Thiazol-4-ylboronic Acid, TRC

Product Code. 30452524
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
missing translation for 'unitSize':
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30452524 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30452524 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT344720500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiazol-4-ylboronic Acid
CAS 1016642-07-1
Molecular Formula C3 H4 B N O2 S
Quantity 500 mg
InChI Formula InChI=1S/C3H4BNO2S/c6-4(7)3-1-8-2-5-3/h1-2,6-7H
SMILES OB(O)c1cscn1
IUPAC Name 1,3-thiazol-4-ylboronic acid
Molecular Weight (g/mol) 128.945
Formula Weight 129.006
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.