missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-4-carboxaldehyde, 95%

Product Code. 15471049
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471049 5 g 5g
15481049 250 mg 250mg
15461049 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471049 Supplier Thermo Scientific Alfa Aesar Supplier No. H64028.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Thiazole-4-carboxaldehyde, 95%, C4H3NOS, CAS Number-3364-80-5, 4-formylthiazole, 4-thiazolecarbaldehyde, 1,3-thiazole-4-carboxaldehyde, 4-formyl-1,3-thiazole, thiazole-4-carbaldehyde, thiazole-4-carboxyaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxaldehyde, pubchem13812, zlchem 1299, 5g | CAS: 3364-80-5 | C4H3NOS | 113.13 g/mol

Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3364-80-5
Molecular Formula C4H3NOS
Molecular Weight (g/mol) 113.13
MDL Number MFCD00626896
InChI Key WRFKSVINLIQRKF-UHFFFAOYSA-N
Synonym thiazole-4-carboxaldehyde, thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxyaldehyde, 4-formylthiazole, 1,3-thiazole-4-carboxaldehyde, 4-thiazolecarbaldehyde, zlchem 1299, pubchem13812, 4-formyl-1,3-thiazole
PubChem CID 2763214
IUPAC Name 1,3-thiazole-4-carbaldehyde
SMILES O=CC1=CSC=N1

Specifications

Melting Point 61°C to 65°C
Color Brown
Quantity 5 g
Beilstein 107137
Solubility Information Slightly soluble in water.
Formula Weight 113.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Thiazole-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.