missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-5-carboxaldehyde, 95%, Thermo Scientific™

Product Code. 15466608
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466608 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15466608 Supplier Thermo Scientific Alfa Aesar Supplier No. H60138.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1003-32-3
Density 1.304
Boiling Point 92°C to 98°C (16 mmHg)
Flash Point 98°C (208°F)
Molecular Formula C4H3NOS
Refractive Index 1.5874
MDL Number MFCD02179516
Quantity 1 g
Synonym thiazole-5-carboxaldehyde, thiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde, thiazole-5-carboxyaldehyde, 5-formylthiazole, 5-formyl-1,3-thiazole, 1,3-thiazole-5-carboxaldehyde, thiazole-5-aldehyde, zlchem 512, pubchem14219
InChI Key ZXRLWHGLEJGMNO-UHFFFAOYSA-N
SMILES O=CC1=CN=CS1
IUPAC Name 1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol) 113.13
PubChem CID 2773577
Formula Weight 113.14
Percent Purity 95%
Chemical Name or Material Thiazole-5-carboxaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.