missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine, 97%

Product Code. 11461390
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461390 1 g 1g
11471390 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461390 Supplier Thermo Scientific Alfa Aesar Supplier No. H26427.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 504-78-9 | C3H8NS | 90.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-78-9
Molecular Formula C3H8NS
Molecular Weight (g/mol) 90.16
MDL Number MFCD00005211
InChI Key OGYGFUAIIOPWQD-UHFFFAOYSA-O
Synonym thiazolidine, tetrahydrothiazole, 1-thia-3-azacyclopentane, thiazole, tetrahydro, unii-i320806bkw, ccris 4275, acmc-1ao0m, 4-27-00-00009 beilstein handbook reference
PubChem CID 10444
ChEBI CHEBI:50120
IUPAC Name 1,3-thiazolidine
SMILES C1CSC[NH2+]1

Specifications

Density 1.131
Boiling Point 165°C
Flash Point 56°C (133°F)
Odor Characteristic
Refractive Index 1.5545
Quantity 1 g
UN Number UN1993
Formula Weight 89.16
Percent Purity 97%
Chemical Name or Material Thiazolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.