missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine, 98%

Product Code. 10571082 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10571082 5 g Glass bottle 5g
10088671 1 g Glass bottle 1g
10110172 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10571082 Supplier Thermo Scientific Acros Supplier No. 165310050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 504-78-9 | C3H8NS | 90.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-78-9
Molecular Formula C3H8NS
Molecular Weight (g/mol) 90.16
MDL Number MFCD00005211
InChI Key OGYGFUAIIOPWQD-UHFFFAOYSA-O
Synonym thiazolidine, tetrahydrothiazole, 1-thia-3-azacyclopentane, thiazole, tetrahydro, unii-i320806bkw, ccris 4275, acmc-1ao0m, 4-27-00-00009 beilstein handbook reference
PubChem CID 10444
ChEBI CHEBI:50120
SMILES C1CSC[NH2+]1

Specifications

Color Colorless
Density 1.1310g/mL
Boiling Point 72°C to 75°C (25.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5525 to 1.5545
Quantity 5 g
Specific Gravity 1.131
IUPAC Name 1,3-thiazolidine
Formula Weight 89.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Thiazolidine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thiazolidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.