missing translation for 'onlineSavingsMsg'
Learn More

Thiazolylacetylglycine Oxime, TRC

Product Code. 30457128
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457128 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457128 Supplier Toronto Research Chemicals Supplier No. TRCT3442305MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiazolylacetylglycine Oxime
CAS 178949-03-6
Molecular Formula C7 H8 N4 O4 S
Quantity 5 mg
Synonym (Z)-N-[(2-Amino-4-thiazolyl)(hydroxyimino)acetyl]glycine, N-[(2Z)-(2-Amino-4-thiazolyl)(hydroxyimino)acetyl]glycine, Cefdinir Thiazolylacetyl Glycine Oxime Impurity (USP)
InChI Formula InChI=1S/C7H8N4O4S/c8-7-10-3(2-16-7)5(11-15)6(14)9-1-4(12)13/h2,15H,1H2,(H2,8,10)(H,9,14)(H,12,13)/b11-5-
SMILES Nc1nc(cs1)\C(=N\O)\C(=O)NCC(=O)O
IUPAC Name 2-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]acetic acid
Molecular Weight (g/mol) 244.23
Formula Weight 244.0266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.