missing translation for 'onlineSavingsMsg'
Learn More

Thieno[2,3-b]pyrazin-7-amine, Thermo Scientific™™

Product Code. 10431504
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10431504 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10431504 Supplier Thermo Scientific Maybridge Supplier No. MO07822DA
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 59944-75-1
Packaging Amber Glass Bottle
Molecular Formula C6H5N3S
MDL Number MFCD08271970
Quantity 1 g
Synonym thieno 2,3-b pyrazin-7-amine, thieno 3,2-b pyrazin-7-amine, 7-aminothieno 2,3-b pyrazine, 4ctk, svn, 7-aminothieno 2,3-b pyrazin, thiopheno 3,2-b pyrazine-7-ylamine
InChI Key ZRVKSPNBHZCQKY-UHFFFAOYSA-N
SMILES C1=CN=C2C(=N1)C(=CS2)N
IUPAC Name thieno[2,3-b]pyrazin-7-amine
Molecular Weight (g/mol) 151.187
PubChem CID 12299130
Formula Weight 151.1858
Chemical Name or Material Thieno[2, 3-b]pyrazin-7-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.