missing translation for 'onlineSavingsMsg'
Learn More

Thieno[2,3-b]pyrazine-6-carboxylic acid, 90%, Thermo Scientific™

Product Code. 10177992
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10177992 1 g Amber Glass Bottle 1g
10167992 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10177992 Supplier Thermo Scientific Maybridge Supplier No. CC31501DA
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 59944-79-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
InChI Key MEHCDDSVVYRWJT-UHFFFAOYSA-N
Synonym thieno 2,3-b pyrazine-6-carboxylic acid, pubchem9882, thieno 2,3-b pyrazine-6-carboxylicacid, 6-thieno 2,3-b pyrazinecarboxylic acid, thieno 3,2-b pyrazine-6-carboxylic acid, thiopheno 2,3-b pyrazine-6-carboxylic acid
PubChem CID 7164578
IUPAC Name thieno[2,3-b]pyrazine-6-carboxylic acid
SMILES C1=CN=C2C(=N1)C=C(S2)C(=O)O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 180.19
Percent Purity 90%
Chemical Name or Material Thieno[2, 3-b]pyrazine-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.