missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide Solution, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11424749 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Tamper evident
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11424749 1 L Tamper evident 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424749 Supplier Reagecon Supplier No. 1089602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to Eur. Pharm. Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Thioacetamide Solution, EP Grade, Reagecon™ is a ready-to-use reagent with 40g/L of C2H5NS in water used to supply sulfide ions in manual and automated titrations.

Thioacetamide is an organosulfur compound that serves as a source of sulfide ions in the synthesis of organic and inorganic compounds. When a solution of thioacetamide is used to treat an aqueous solution of many metal cations, the corresponding metal sulfide results. This solution is prepared by adding 40g per 1L water.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions

Specifications

CAS 62-55-5, 7732-18-5
Color Colorless
Odor Slight mercaptan
Packaging Tamper evident
Molecular Formula C2H5NS
MDL Number MFCD00008070
Quantity 1 L
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
SMILES CC(N)=S
IUPAC Name ethanethioamide
Molecular Weight (g/mol) 75.13
PubChem CID 2723949
ChEBI CHEBI:32497
Formula Weight 75.1g/mol
Grade Eu. Ph. Chapter 4(4.1.1)
Concentration or Composition (by Analyte or Components) 40g/L
Physical Form Liquid
Chemical Name or Material Thioacetamide
Show More Show Less
compliance-icons
Product Identifier
  • THIOACETAMIDE SOLUTION
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : thioacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.