missing translation for 'onlineSavingsMsg'
Learn More

Thioacetic acid, 98%

Product Code. 10074043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10074043 2.5 kg Glass bottle 2.5kg
10571381 100 g Glass bottle 100g
10081650 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10074043 Supplier Thermo Scientific Acros Supplier No. 138780025
  • ONLINE EXCLUSIVE PRICE
    555.00 €
    538.00 € / 2.5kg
    Save 17.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 507-09-5 | C2H4OS | 76.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-09-5
Molecular Formula C2H4OS
Molecular Weight (g/mol) 76.11
MDL Number MFCD00004853
InChI Key DUYAAUVXQSMXQP-UHFFFAOYSA-N
Synonym thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
PubChem CID 10484
ChEBI CHEBI:16555
IUPAC Name ethanethioic S-acid
SMILES CC(S)=O

Specifications

Melting Point -17.0°C
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 87.0°C to 93.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3COSH
Refractive Index 1.4630 to 1.4650
Quantity 2.5 kg
Beilstein 02, 230
Fieser 01,1154
Merck Index 15, 9473
Specific Gravity 1.07
Solubility Information Solubility in water: 27g/L water (15°C) (hydrolyse). Other solubilities: soluble in alcohol and ether
Formula Weight 76.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Thioacetic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.