missing translation for 'onlineSavingsMsg'
Learn More

Thioacetic acid, 98%

Product Code. 10081650 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10081650

Brand: Thermo Scientific Acros 138785000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 507-09-5 | C2H4OS | 76.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-09-5
Molecular Formula C2H4OS
Molecular Weight (g/mol) 76.11
MDL Number MFCD00004853
InChI Key DUYAAUVXQSMXQP-UHFFFAOYSA-N
Synonym thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
PubChem CID 10484
ChEBI CHEBI:16555
IUPAC Name ethanethioic S-acid
SMILES CC(S)=O

Specifications

Melting Point -17.0°C
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 87.0°C to 93.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3COSH
Refractive Index 1.4630 to 1.4650
Quantity 500 g
Beilstein 02, 230
Fieser 01,1154
Merck Index 15, 9473
Specific Gravity 1.07
Solubility Information Solubility in water: 27g/L water (15°C) (hydrolyse). Other solubilities: soluble in alcohol and ether
Formula Weight 76.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Thioacetic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.