missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzamide, 95%

Product Code. 10781241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10781241 50 g 50g
10113990 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10781241 Supplier Thermo Scientific Acros Supplier No. 164730500
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2227-79-4 | C7H7NS | 137.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2227-79-4
Molecular Formula C7H7NS
Molecular Weight (g/mol) 137.20
MDL Number MFCD00008060
InChI Key QIOZLISABUUKJY-UHFFFAOYSA-N
Synonym thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione
PubChem CID 683563
ChEBI CHEBI:80418
IUPAC Name benzenecarbothioamide
SMILES NC(=S)C1=CC=CC=C1

Specifications

Melting Point 114°C to 118°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CSNH2
Quantity 50 g
Beilstein 09, 424
Solubility Information Solubility in water: slightly soluble
Formula Weight 137.21
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Thiobenzamide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Thiobenzamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.