missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzamide, 98%

Product Code. 11419843
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419843 25 g 25g
11429843 100 g 100g
11439843 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419843 Supplier Thermo Scientific Alfa Aesar Supplier No. A12973.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2227-79-4 | C7H7NS | 137.20 g/mol

Thiobenzamide is useful for the preparation of amide and amidine adducts. It is also used to prepare 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides. Further, it acts as a raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiobenzamide is useful for the preparation of amide and amidine adducts. It is also used to prepare 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides. Further, it acts as a raw material in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2227-79-4
Molecular Formula C7H7NS
Molecular Weight (g/mol) 137.20
MDL Number MFCD00008060
InChI Key QIOZLISABUUKJY-UHFFFAOYSA-N
Synonym thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione
PubChem CID 683563
ChEBI CHEBI:80418
IUPAC Name benzenecarbothioamide
SMILES NC(=S)C1=CC=CC=C1

Specifications

Melting Point 114°C to 118°C
Flash Point 120°C (248°F)
Quantity 25 g
UN Number UN2811
Beilstein 606021
Solubility Information Slightly soluble in water.
Formula Weight 137.21
Percent Purity 98%
Chemical Name or Material Thiobenzamide

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.