missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzoic Acid (94%)(Benzenecarbothioic Acid), TRC

Product Code. 30339713
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339713 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339713 Supplier Toronto Research Chemicals Supplier No. TRCB189000100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiobenzoic Acid
CAS 98-91-9
Molecular Formula C7H6OS
Quantity 100 g
Synonym Benzoyl Thiol, Monothiobenzoic Acid, NSC 66502, Benzenecarbothioic Acid
InChI Formula InChI=1S/C7H6OS/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)
SMILES SC(=O)c1ccccc1
IUPAC Name benzenecarbothioic S-acid
Molecular Weight (g/mol) 138.19
Formula Weight 138.0139
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.