missing translation for 'onlineSavingsMsg'
Learn More

Thiodiglycolic acid, 98%

Product Code. 11375139
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375139 25 g 25g
11385139 100 g 100g
11395139 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375139 Supplier Thermo Scientific Alfa Aesar Supplier No. A17880.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 123-93-3 | C4H6O4S | 150.148 g/mol

It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.

Solubility
Solubility in water is almost transparent. (water, 4.00E+05 mg/L @ 25°C (exp); 1e+006 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-93-3
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.148
MDL Number MFCD00004359
InChI Key UVZICZIVKIMRNE-UHFFFAOYSA-N
Synonym thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
PubChem CID 31277
ChEBI CHEBI:35012
IUPAC Name 2-(carboxymethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCC(=O)O

Specifications

Melting Point 127°C to 131°C
UN Number UN3261
Beilstein 1764392
Merck Index 149331
Formula Weight 150.15
Percent Purity 98%
Quantity 25 g
Chemical Name or Material Thiodiglycolic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.