missing translation for 'onlineSavingsMsg'
Learn More

Thiodiglycolic Anhydride, TRC

Product Code. 30453273
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453273 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453273 Supplier Toronto Research Chemicals Supplier No. TRCT34452525G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiodiglycolic Anhydride
CAS 3261-87-8
Molecular Formula C4H4O3S
Quantity 25 g
Synonym 1,4-Oxathiane-2,6-dione, Thiodiacetic Acid Cyclic Anhydride, 2,2'-Thiodiglycolic Anhydride, 2,2'-Thiobisacetic Acid Cyclic Anhydride, NSC 147625
InChI Formula InChI=1S/C4H4O3S/c5-3-1-8-2-4(6)7-3/h1-2H2
SMILES O=C1CSCC(=O)O1
IUPAC Name 1,4-oxathiane-2,6-dione
Molecular Weight (g/mol) 132.14
Formula Weight 131.9881
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.