missing translation for 'onlineSavingsMsg'
Learn More

Thioglycolic Acid 2-Ethylhexyl Ester, TRC

Product Code. 30008903
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
Unit Size:
10mL
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008903 1 mL 1mL
30006510 10 mL 10mL
30004033 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008903 Supplier Toronto Research Chemicals Supplier No. TRCT3507631ML
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7659-86-1
Molecular Formula C10H20O2S
Molecular Weight (g/mol) 204.33
Synonym 2-Ethylhexyl 2-Mercaptoacetate, 2-Ethylhexyl 2-Sulfanylacetate, 2-Ethylhexyl Mercaptoacetate, 2-Ethylhexyl Thioglycolate, 2-Mercaptoacetic Acid 2-Ethylhexyl Ester, EHTG, Mercaptoacetic Acid 2-Ethylhexyl Ester;
IUPAC Name 2-ethylhexyl 2-sulfanylacetate
SMILES CCCCC(CC)COC(=O)CS

Specifications

Quantity 1 mL
InChI Formula InChI=1S/C10H20O2S/c1-3-5-6-9(4-2)7-12-10(11)8-13/h9,13H,3-8H2,1-2H3
Formula Weight 204.1184
Chemical Name or Material Thioglycolic Acid 2-Ethylhexyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.