missing translation for 'onlineSavingsMsg'
Learn More

Thioglycolic acid, 99.94%, Bacteriological grade, MP Biomedicals™
BIOPHARMA

Product Code. 11436342 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436342 100 g 100g
11426342 25 g 25g
11446342 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11436342 missing translation for 'mfr' MP Biomedicals missing translation for 'supplierNo' 0210293380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 367-51-1
Molecular Formula C2H3NaO2S
Molecular Weight (g/mol) 114.094
InChI Key GNBVPFITFYNRCN-UHFFFAOYSA-M
Synonym sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
PubChem CID 23690444
ChEBI CHEBI:86481
IUPAC Name sodium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[Na+]

Specifications

Color White
Quantity 100 g
Formula Weight 114.11
Percent Purity 0.9994
Grade Bacteriological
Physical Form Crystalline Powder
Chemical Name or Material Thioglycolic acid
compliance-icons
Product Identifier
  • Thioglycolic Acid Sodium Salt, biological grade
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Thioglycolic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.