missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ ThiolTracker™ Violet (glutathione detection reagent)

Product Code. 10734274
Change view
Click to view available options
Quantity:
180 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10734274 180 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10734274 Supplier Molecular Probes™ Supplier No. T10095
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

ThiolTracker™ Violet (glutathione detection reagent)

ThiolTracker™ Violet is a brighter and more robust intracellular thiol probe for reproducible GSH detection. Since reduced glutathione represents the majority of intracellular free thiols in the cell, ThiolTracker™ Violet can be used in estimating the cellular level of reduced glutathione.
ThiolTracker™ Violet offers the following;
  • 10X brighter than mBCL
  • UV or 405 nm excitation, 525 nm emission
  • Easy to use
  • Reproducible results
  • Can be fixed with aldehydes and permeabilized by Triton™ X-100 (0.5%)
  • Formaldehyde fixable and suvives detergent extraction
  • Can be used in multiplex assays including cytotoxicity studies
    Subcellular detection and localization of GSH is important in understanding the modulation of redox status, the effect of drugs, and the mechanisms of detoxification. Furthermore, differences in GSH levels in response to oxidative stress in subpopulations of cells have been reported, underscoring the importance of detection methods amenable to flow cytometry and automated fluorescence detection. ThiolTracker™ Violet can be used for Flow Cytometry, HCS imaging, and epifluorescent microscopy.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Color Violet
    Content And Storage

    Store at ≤–20°C, L protect material from long-term exposure to light; may be exposed to light for short periods of time, requires DMSO

    Excitation Wavelength Range 405⁄526
    Dye Type ThiolTracker™ Violet
    Format Tube(s), Slide(s)
    For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
    Quantity 180 reactions
    Detection Method Fluorescence
    Emission Visible
    Form Lyophilized
    Product Line Molecular Probes, ThiolTracker
    Shipping Condition Room Temperature
    Label Type Other Labels or Dyes
    Product Type Actin
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.