missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine 1,1-dioxide, Thermo Scientific Chemicals

Product Code. 17211192 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17211192 25 g 25g
17201192 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17211192 Supplier Thermo Scientific Acros Supplier No. 466030250
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39093-93-1 | C4H9NO2S | 135.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39093-93-1
Molecular Formula C4H9NO2S
Molecular Weight (g/mol) 135.18
InChI Key NDOVLWQBFFJETK-UHFFFAOYSA-N
IUPAC Name 1λ⁶-thiomorpholine-1,1-dione
SMILES O=S1(=O)CCNCC1

Specifications

Melting Point 68.0°C to 72.0°C
Quantity 25 g
Formula Weight 135.18
Chemical Name or Material Thiomorpholine 1,1-dioxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.