missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine, 97%

Product Code. 10522614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10522614 25 g Glass bottle 25g
10452855 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10522614 Supplier Thermo Scientific Acros Supplier No. 187970250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-90-0 | C4H9NS | 103.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-90-0
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.19
MDL Number MFCD00005974
InChI Key BRNULMACUQOKMR-UHFFFAOYSA-N
Synonym thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
PubChem CID 67164
ChEBI CHEBI:36392
IUPAC Name thiomorpholine
SMILES C1CSCCN1

Specifications

Density 1.0880g/mL
Boiling Point 175°C to 178°C
Flash Point 63°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5370 to 1.539
Quantity 25 g
Beilstein 27, 9
Merck Index 15, 9452
Specific Gravity 1.088
Solubility Information Solubility in water: miscible. Other solubilities: miscible with many organic liquids
Formula Weight 103.19
Percent Purity 97%
Chemical Name or Material Thiomorpholine
Show More Show Less
compliance-icons
Product Identifier
  • Thiomorpholine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.