missing translation for 'onlineSavingsMsg'
Learn More

Thionicotinamide, 99%

Product Code. 10346830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
Product Code. Quantity Packaging unitSize
10346830 100 g Glass bottle 100g
10255330 25 g Glass bottle 25g
2 options
This item is not returnable. View return policy

Product Code. 10346830

Brand: Thermo Scientific Acros 138801000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4621-66-3 | C6H6N2S | 138.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4621-66-3
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.19
MDL Number MFCD00006399
InChI Key XQWBMZWDJAZPPX-UHFFFAOYSA-N
Synonym thionicotinamide, 3-pyridinecarbothioamide, nicotinothioamide, 3-thiocarbamoylpyridine, 3-thioamidopyridine, nicotinamide, thio, 3-pyridylthioformamide, 3-pyridinethiocarboxamide, thio-3-pyridinecarboxamide, unii-6a0hxx0753
PubChem CID 737155
IUPAC Name pyridine-3-carbothioamide
SMILES NC(=S)C1=CC=CN=C1

Specifications

Melting Point 185°C to 190°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: 0.2g/100 mL (20°C)
Formula Weight 138.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Thionicotinamide, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.