missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thionin acetate, pure, high purity biological stain

Product Code. 10444345 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444345 25 g Glass bottle 25g
10033030 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10444345 Supplier Thermo Scientific Chemicals Supplier No. 229840250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78338-22-4 | C14H13N3O2S | 287.337 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78338-22-4
Molecular Formula C14H13N3O2S
Molecular Weight (g/mol) 287.337
MDL Number MFCD00081194
InChI Key JOIRQYHDJINFGA-UHFFFAOYSA-N
Synonym 3, 7-Diaminophenothiazin-5-ium acetate
PubChem CID 2724414
IUPAC Name (7-aminophenothiazin-3-ylidene)azanium;acetate
SMILES CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2

Specifications

Dye Content 85% min.
Beilstein 27, 392
Merck Index 14, 9346
Solubility Information Solubility in water: ca 2.5g/L (25°C). Other solubilities: practically insoluble in ethanol
Infrared Spectrum Authentic
Lambda Max 597 to 601nm (in water)
Formula Weight 287.33
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Green
Quantity 25 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Thionin acetate, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Thionin acetate, high purity biological stain
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Thionin acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.