missing translation for 'onlineSavingsMsg'
Learn More

Thionyl bromide, 97%

Product Code. 11362727
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11362727 100 g 100g
11352727 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11362727

Brand: Thermo Scientific Alfa Aesar L15648.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 507-16-4 | Br2OS | 207.87 g/mol

Thionyl bromide is used as a brominating agent in organic synthesis. It is used in the bromination of alfa-beta-unsaturated carbonyl compounds. It is also used to prepare alkyl bromides from alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thionyl bromide is used as a brominating agent in organic synthesis. It is used in the bromination of alfa-beta-unsaturated carbonyl compounds. It is also used to prepare alkyl bromides from alcohols.

Solubility
Miscible with benzene, toluene and ether.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, hydrobromide, acetone and alcohol.It hydrolyzes readily to release hydrogen bromide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-16-4
Molecular Formula Br2OS
Molecular Weight (g/mol) 207.87
MDL Number MFCD00011448
InChI Key HFRXJVQOXRXOPP-UHFFFAOYSA-N
Synonym thionyl bromide, thionyl dibromide, unii-n6cu78b13t, thionylbromide, sulfur oxy dibromide, acmc-20ajin, pubchem2023, sobr2, thionyl bromide, purum at
PubChem CID 68176
IUPAC Name sulfurooyl dibromide
SMILES BrS(Br)=O

Specifications

Boiling Point 47°C to 49°C (20mmHg)
Melting Point -52°C
Quantity 100 g
UN Number UN3264
Merck Index 14,9347
Solubility Information Miscible with benzene,toluene and ether.
Formula Weight 207.88
Percent Purity 97%
Odor Acrid
Refractive Index 1.675
Sensitivity Light and moisture sensitive
Density 2.683
Chemical Name or Material Thionyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.