missing translation for 'onlineSavingsMsg'
Learn More

Thiopalmitic Acid (~90%), TRC

Product Code. 30456412
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456412 25 mg 25mg
30451969 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456412 Supplier Toronto Research Chemicals Supplier No. TRCT36807025MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7530-93-0
Molecular Formula C16H32OS
Molecular Weight (g/mol) 272.49
Synonym Hexadecanethioic Acid
IUPAC Name hexadecanethioic O-acid
SMILES CCCCCCCCCCCCCCCC(=S)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H32OS/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)
Formula Weight 272.2174
Chemical Name or Material Thiopalmitic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.