missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2,5-dicarboxylic acid, 97%

Product Code. 11398345
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398345 5 g 5g
11388345 1 g 1g
11308355 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398345 Supplier Thermo Scientific Alfa Aesar Supplier No. L10293.06
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4282-31-9 | C6H4O4S | 172.154 g/mol

Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4282-31-9
Molecular Formula C6H4O4S
Molecular Weight (g/mol) 172.154
MDL Number MFCD00016896
InChI Key YCGAZNXXGKTASZ-UHFFFAOYSA-N
Synonym 2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
PubChem CID 20273
IUPAC Name thiophene-2,5-dicarboxylic acid
SMILES C1=C(SC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 355°C to 360°C
Quantity 5 g
Beilstein 136781
Solubility Information Very soluble in water.
Formula Weight 172.16
Percent Purity 97%
Chemical Name or Material Thiophene-2,5-dicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.