missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-boronic acid pinacol ester, 98%

Product Code. 15430719
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430719 1 g 1g
15440719 5 g 5g
15450719 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430719 Supplier Thermo Scientific Alfa Aesar Supplier No. H63819.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 193978-23-3 | C10H15BO2S | 210.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193978-23-3
Molecular Formula C10H15BO2S
Molecular Weight (g/mol) 210.098
MDL Number MFCD05663878
InChI Key FFZHICFAHSDFKZ-UHFFFAOYSA-N
Synonym 2-thiopheneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl-1,3,2-dioxaborolane, thiophene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene, thiophene-2-boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-thienyl, 4,4,5,5-tetramethyl-2-thien-2-yl-1,3,2-dioxaborolane, pubchem18441
PubChem CID 10703628
IUPAC Name 4,4,5,5-tetramethyl-2-thiophen-2-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CS2

Specifications

Melting Point 65°C to 70°C
Quantity 1 g
Formula Weight 228.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Thiophene-2-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.