missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxamide, 99%

Product Code. 11422248
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422248 5 g 5g
11432248 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422248 Supplier Thermo Scientific Alfa Aesar Supplier No. A15219.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5813-89-8 | C5H5NOS | 127.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5813-89-8
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00010425
InChI Key DENPQNAWGQXKCU-UHFFFAOYSA-N
Synonym 2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference
PubChem CID 22063
IUPAC Name thiophene-2-carboxamide
SMILES C1=CSC(=C1)C(=O)N

Specifications

Melting Point 178°C to 183°C
Quantity 5 g
Beilstein 110153
Formula Weight 127.17
Percent Purity 99%
Chemical Name or Material Thiophene-2-carboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.