missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carboxylic hydrazide, 97%

Product Code. 11463624
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463624 25 g 25g
11453624 5 g 5g
11473624 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463624 Supplier Thermo Scientific Alfa Aesar Supplier No. A11270.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2361-27-5 | C5H6N2OS | 142.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2361-27-5
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.176
MDL Number MFCD00005435
InChI Key SOGBOGBTIKMGFS-UHFFFAOYSA-N
Synonym 2-thiophenecarboxylic acid hydrazide, thiophene-2-carboxylic acid hydrazide, 2-thenoylhydrazine, 2-thiophenecarboxylic hydrazide, 2-thiophenecarbohydrazide, thiophene-2-carboxylic hydrazide, 2-thiophenecarboxylic acid, hydrazide, 2-thiophenecarbohydrazonic acid, 2-thienylcarboxylic acid hydrazide, 2-thiophene carboxylic hydrazide
PubChem CID 73098
IUPAC Name thiophene-2-carbohydrazide
SMILES C1=CSC(=C1)C(=O)NN

Specifications

Melting Point 137°C to 140°C
Quantity 25 g
Beilstein 114437
Formula Weight 142.18
Percent Purity ≥98%
Chemical Name or Material Thiophene-2-carboxylic hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.