missing translation for 'onlineSavingsMsg'
Learn More

Thioridazine 5-Sulfoxide (Mixture of Diastereomers), TRC

Product Code. 30135722
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135722 50 mg 50mg
30004813 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135722 Supplier Toronto Research Chemicals Supplier No. TRCT36881550MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7776-05-8
Molecular Formula C21 H26 N2 O S2
Molecular Weight (g/mol) 386.57
Synonym 10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulfanyl)-10H-phenothiazine 5-oxide, 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylthio)-10H-phenothiazine 5-oxide, Phenothiazine, 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylthio)-, 5-oxide (7CI,8CI), Thioridazine 5-oxide, Thioridazine 5-sulfoxide, Thioridazine sulfoxide
IUPAC Name 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfanylphenothiazine 5-oxide
SMILES CSc1ccc2c(c1)N(CCC3CCCCN3C)c4ccccc4S2=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H26N2OS2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)26(24)21-11-10-17(25-2)15-19(21)23/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3
Formula Weight 386.1487
Chemical Name or Material Thioridazine 5-Sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.