missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Thioridazine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Thioridazine-d3 Hydrochloride |
| CAS | 1189928-36-6 |
| Molecular Formula | C21 D3 H23 N2 S2 . Cl H |
| Quantity | 0.01 g |
| Synonym | 2-Methylsulfanyl-10-[2-[1-(trideuteriomethyl)piperidin-2-yl]ethyl]phenothiazine hydrochloride, 10-[2-[1-(Methyl-d3)piperidin-2-yl]ethyl]-2-(methylsulfanyl)-10 H-phenothiazine hydrochloride (1:1), 2-Methylmercapto-10-[2-[N-(methyl-d3)-2-piperidyl]ethyl]phenothiazine hydrochloride, Thioridazine-d3 Hydrochloride, Thioridazine-D3 (N-methyl-D3) Hydrochloride, (±)-Thioridazine-d3 HCl (N-methyl-d3) |
| InChI Formula | InChI=1 S/C21H26N2S2.ClH/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(24-2)15-19(21)23;/h3-4,8-11,15-16 H,5-7,12-14H2,1-2H3;1 H/i1D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N1CCCCC1CCN2c3ccccc3Sc4ccc(SC)cc24 |
| IUPAC Name | 2-methylsulfanyl-10-[2-[1-(trideuteriomethyl)piperidin-2-yl]ethyl]phenothiazine;hydrochloride |
| Molecular Weight (g/mol) | 410.054 |
| Formula Weight | 409.149 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction