missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Thioridazine hydrochloride
Description
Dopamine receptor antagonist that displays antipsychotic activity. Exhibits anticancer activity in cervical, endometrial and breast cancer cells; triggers apoptosis by targeting the PI 3-K/Akt/mTOR/p70 S6K pathway. Induces G1 cell cycle arrest. Selective inducer of cancer stem cell (CSC) differentiation.
Specifications
Specifications
| CAS | 130-61-0 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Molecular Formula | C21H27ClN2S2 |
| Formula Weight | 407.04 |
| Purity | >99% |
| Molecular Weight (g/mol) | 407.031 |
| Chemical Name or Material | Thioridazine hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?