missing translation for 'onlineSavingsMsg'
Learn More

Thiosalicylic acid, 98%

Product Code. 11475413
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475413 50 g 50g
11485413 250 g 250g
11465413 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475413 Supplier Thermo Scientific Alfa Aesar Supplier No. A13401.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 147-93-3 | C7H6O2S | 154.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Thiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.

Solubility
Soluble in dimethyl sulfoxide, alcohol and ether. Slightly soluble in water, ethanol and alkanes.

Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-93-3
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00004836
InChI Key NBOMNTLFRHMDEZ-UHFFFAOYSA-N
Synonym thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
PubChem CID 5443
ChEBI CHEBI:59124
IUPAC Name 2-sulfanylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)S

Especificaciones

Melting Point 165°C to 168°C
Merck Index 149360
Solubility Information Soluble in dimethyl sulfoxide,alcohol and ether. Slightly soluble in water,ethanol and alkanes.
Formula Weight 154.19
Percent Purity 98%
Density 1.49
Quantity 50 g
Odor Stench
Sensitivity Air and light sensitive
Chemical Name or Material Thiosalicylic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.