missing translation for 'onlineSavingsMsg'
Learn More

Thiosemicarbazide, 98%

Product Code. p-7022044
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417586 100 g 100g
11427586 500 g 500g
11437586 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417586 Supplier Thermo Scientific Alfa Aesar Supplier No. A14630.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-19-6 | CH5N3S | 91.132 g/mol

Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.

Solubility
Soluble in water, ethanol, alcohols and hot methanol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-19-6
Molecular Formula CH5N3S
Molecular Weight (g/mol) 91.132
MDL Number MFCD00007620
InChI Key BRWIZMBXBAOCCF-UHFFFAOYSA-N
Synonym thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine
PubChem CID 2723789
IUPAC Name aminothiourea
SMILES C(=S)(N)NN

Specifications

Melting Point ∼180°C (decomposition)
Odor Odorless
Quantity 100 g
UN Number UN2811
Beilstein 506320
Merck Index 14,9361
Solubility Information Soluble in water,ethanol,alcohols and hot methanol.
Formula Weight 91.14
Percent Purity 99%
Chemical Name or Material Thiosemicarbazide
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.