missing translation for 'onlineSavingsMsg'
Learn More

Thiourea, 99%, extra pure

Product Code. 10785971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
Unit Size:
1kg
25kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10785971 1 kg Plastic bottle 1kg
10497140 500 g Plastic bottle 500g
10338703 5 kg Plastic drum 5kg
15199183 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10785971 Supplier Thermo Scientific Acros Supplier No. 138910010
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thiourea, Thiocarbamide, CAS # 62-56-6, is an analogue of urea with a single sulfur-substituted oxygen. | CAS: 62-56-6 | CH4N2S | 76.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Thiourea is a derivative of the biological compound, urea
• Thiourea is highly toxic when it decomposes and acts as a carcinogen

Applications

• Thiourea can be used as a denaturant in lysis buffers and similar sample buffers that require solubilization of proteins
• Thiourea can act as an antioxidant and chromophore
• Thiourea can be used in the study of cellular redox, autophagy, and aging

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-56-6
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00008067
InChI Key UMGDCJDMYOKAJW-UHFFFAOYSA-N
Synonym thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
PubChem CID 2723790
ChEBI CHEBI:36946
IUPAC Name thiourea
SMILES NC(N)=S

Specifications

Melting Point 171.0°C to 175.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodimetry)
Packaging Plastic bottle
Linear Formula H2NCSNH2
Quantity 1 kg
Beilstein 03, 180
Fieser 01,1164; 02,412; 03,290; 06,586; 11,519; 15,308
Merck Index 15, 9521
Solubility Information Solubility in water: 136g/L (20°C). Other solubilities: 12g/100g methanol (20°C),soluble in isopropylalcohol,butanol ,pentanol,sparingly soluble in ether
Formula Weight 76.12
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Thiourea
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thiourea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.