missing translation for 'onlineSavingsMsg'
Learn More

Thiourea, ACS, 99% min.

Product Code. 11368048
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368048 50 g 50g
11358048 250 g 250g
11378048 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368048 Supplier Thermo Scientific Alfa Aesar Supplier No. 036609.18
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent for organic synthesis | CAS: 62-56-6 | CH4N2S | 76.12 g/mol

Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-56-6
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00008067
InChI Key UMGDCJDMYOKAJW-UHFFFAOYSA-N
Synonym thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
PubChem CID 2723790
ChEBI CHEBI:36946
IUPAC Name thiourea
SMILES NC(N)=S

Specifications

Melting Point 175°C to 178°C
Color White
Density 1.405 g/mL
Odor Odorless
Linear Formula H2NCSNH2
Quantity 50 g
UN Number UN3077
Beilstein 605327
Merck Index 14,9367
Solubility Information Soluble in water.
Formula Weight 76.12
Percent Purity ≤99%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Thiourea
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thiourea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.