missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieve Type 3A, Technical, 1.6-2.5mm Beads (8-12 Mesh), Fisher Chemical
BIOPHARMA

Product Code. 12916213 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12916213 250 g 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12916213 Supplier Fisher Chemical Supplier No. MS/1010/50
  • ONLINE EXCLUSIVE PRICE
    279.00 €
    276.08 € / 50mL
    Save 2.92 €
    1% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Thirty three elements solution - Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr,Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Nb, Ni, Pb,Rb Sb Se Sr, Ti, Tl, V, U, Zn for ICP 100mg/L HNO3 matrix

Specifications

Color Beige
Packaging Plastic powder jar
Quantity 250 g
CAS 308080-99-1
Physical Form Solid
Molecular Formula AlNaO6Si2
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
IUPAC Name Molecular sieves 3A
Molecular Weight (g/mol) 202.135
PubChem CID 19758701
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Molecular sieve type 3A Nominal pore size 3 Angstroms
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH207-Warning! Contains cadmium. Dangerous fumes are formed during use. See information supplied by the manufacturer. Comply with the safety instructions.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid, beryllium nitrate, aluminium nitrate, lithium nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.