Learn More
Euro Protection™ Thor Multirisk Pants, Navy
Product Code. 15810892
Change view
Click to view available options
Size:
Small
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15810892 Large Each
15830892 Small Each
15820892 - Each
15840892 X-Large Each
15850892 2X-Large Each
15860892 3X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15810892 Supplier Euro Protection™ Supplier No. 8MTHTNL
  • ONLINE EXCLUSIVE PRICE
    139.65 €
    69.46 € / Pack of 1
    Save 70.19 €
    50% Off
Estimated Shipment: 08-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Euro Protection™ Thor Multirisk Pants, Navy are antistatic, fire-retardant, and feature reflective band tape for visibility.
  • Fire retardant
  • Antistatic
  • Reflective band tape
  • Cotton/polyester construction
Specifications
Size Large
Product Type Multirisk Pants
Material Cotton, Polyester
Resistant To Fire-retardant, Anti-static
Certifications/Compliance (Europe) EN ISO 11611:2007Class 1 - A1IEC 61482-2:2009 Class 1 - 4 kAEN 13034:2005+A1:2009Type PB6EN 1149-5:200N 14404:2004+A1:2010Type 2
Style Includes reflective band tape