Learn More
SCAT™ Polypropylene Thread Adapters
Shop All SCAT Europe Products
Click to view available options
Closure Size:
Thread 1: GL/S40 (f), Thread 2: GL45 (m)
Thread 1: S 60/61 (f) Thread 2: GL45 (m)
Thread 1: S51 (f), Thread 2: GL45 (m)
Thread 1: S55 (f), Thread 2: GL45 (m)
Thread 1: S60/61, Thread 2: S55 (m)
Closure Color:
Colorless
Unit Size:
Each
Beskrivelse
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polypropylene (PP) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Tekniske data
Tekniske data
| Closure Size | Thread 1: S 60/61 (f) Thread 2: GL45 (m) |
| Closure Color | Colorless |
| Liner Material | Polypropylene |
| Product Type | Thread Adapter |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion