Learn More
SCAT™ PTFE Thread Adapters
Product Code. 10120830 Shop All SCAT Europe Products
Change view
Click to view available options
Closure Size:
GL/S40 (f) to GL45 (m)
GL/S40 (f) to S55 (m)
GL32 (f) to GL45 (m)
GL38 (f) to GL45 (m)
GL45 (f) to S55 (m)
GL45 (m) to 2 in. Tri-sure (m)
GL45 (m) to NS29/32 mm (m)
GL45 (m) to R2 in. BSP/G2 in. (m)
GPI 38-23 (f) to GL45 (m)
ND9
S51 (f) to S55 (m)
S60/61 (m) to B83 (f)
UNF 1/4 in. 28 G (f) to GL14 (m)
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size unitSize
10120830 GL38 (f) to GL45 (m) Each
10374900 GL32 (f) to GL45 (m) Each
10318729 GL45 (m) to 2 in. Tri-sure (m) Each
10829840 GL45 (m) to R2 in. BSP/G2 in. (m) Each
10840411 GL45 (f) to S55 (m) Each
10860391 GL45 (m) to NS29/32 mm (m) Each
10384420 GL/S40 (f) to GL45 (m) Each
10705802 GL/S40 (f) to S55 (m) Each
10881051 S51 (f) to S55 (m) Each
10584448 UNF 1/4 in. 28 G (f) to GL14 (m) Each
10130200 GPI 38-23 (f) to GL45 (m) Each
10671211 ND9 Each
17917270 S60/61 (m) to B83 (f) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10120830 Supplier SCAT™ Supplier No. 107992
  • ONLINE EXCLUSIVE PRICE
    71.90 €
    65.87 € / Pack of 1
    Save 6.03 €
    8% Off
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
SCAT™ Safety Caps and SCAT™ Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polytetrafluoroethylene (PTFE) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Closure Size GL38 (f) to GL45 (m)
For Use With (Equipment) Capillary Connector
Material PTFE
Product Type Thread Adapter