Learn More
Buerkle™ Thread Adapters PP
Product Code. 31188944 Shop All Bürkle Products
Click to view available options
Color:
Black
Black, Orange
Colorless
Gray
Violet
Product Code. 31188944
Brand: Buerkle™ 06001008
Polypropylene Thread Adapter facilitates the secure connection of pumps and discharge cocks to a variety of containers.
  • Constructed of polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Material Polypropylene
Thread Size External: S61x6 (DIN61), Internal: G2 in.
Color Gray
Product Type Thread Adapter