Learn More
Bohlender™ Threaded Adaptor
Product Code. 10707522
Click to view available options
Material:
PTFE
Unit Size:
Each
Product Code. 10707522
Brand: Bohlender™ H97840
Only null left
Add to basket
Add to basket
Specifications
For Use With (Equipment) Bottles with GL 32 thread, for GL/S 40 bottle thread, GL 45 top thread
Quantity 1Each
Product Type Threaded Adaptor
Material PTFE