Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key, threaded, hollow
Product Code. 15894528 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24, 29.2 NS
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size unitSize
15894528 21.5 NS Pack of 100
15844528 12.5 NS Pack of 100
15804538 24, 29.2 NS Pack of 100
15874528 18.8 NS Pack of 100
12848041 14.5 NS Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15894528 Supplier DWK Life Sciences Supplier No. 286046004
  • ONLINE EXCLUSIVE PRICE
    178.61 €
    165.90 € / Pack of 100
    Save 12.71 €
    7% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
For securing the hollow DURAN™ tap plug in the ground DURAN™ single way stopcock.
The 3-part tap safety device holds and secures the hollow DURAN™ tap plug in the tap sleeve.
  • 3-part, for hollow tap plugs
  • Three part retaining device holds and secures the tap plug in the tap sleeve
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3), Polypropylene
Autoclavable Autoclavable
Closure Size 21.5 NS
No. per Pack 100/Pk.
For Use With (Application) For securing the hollow DURAN™ tap plug in the ground DURAN™ one-way tap