missing translation for 'onlineSavingsMsg'
Learn More

Abcam Threonine Assay Kit, Fluorescent, 100 Tests

Product Code. 32120792
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120792 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120792 Supplier Abcam Supplier No. ab239726100Test
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fluorescence-based assay kit for measuring threonine levels in cell and tissue extracts, serum, plasma, and biofluids.

  • Detection method: Fluorescent
  • Sample types: Tissue Homogenate, Cell Lysate, Serum, Plasma, Cerebral Spinal Fluid
  • Equipment: Microplate reader
  • Citations: Cited in > 2 publications

Specifications

Detection Location In-Solution Detection
For Use With (Equipment) Microplate Reader
Format Kit
No. of Reactions 100 Tests
Target Molecule Threonine
Detection Method Fluorescence
Sample Type Tissue Homogenate, Cell Lysate, Serum, Plasma, Cerebral Spinal Fluid
Quantity 100 Tests
For Use With (Application) Threonine detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.