missing translation for 'onlineSavingsMsg'
Learn More

Thulium foil, 0.25mm (0.01in) thick, 99.9% (REO)

Product Code. 11345828
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345828 25 x 25 mm 25mm
11365779 50 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345828 Supplier Thermo Scientific Alfa Aesar Supplier No. 010375.FF
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7440-30-4 | Tm | 168.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-30-4
Molecular Formula Tm
Molecular Weight (g/mol) 168.93
MDL Number MFCD00011281
InChI Key FRNOGLGSGLTDKL-UHFFFAOYSA-N
Synonym tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
PubChem CID 23961
ChEBI CHEBI:33380
IUPAC Name thulium
SMILES [Tm]

Specifications

Color Silver to Gray
Quantity 25 x 25 mm
Odor Odorless
Weight ≈1.75g/25x25mm
Form Foil
UN Number UN3178
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Thulium foil, 0.25mm (0.01 in.) thick

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.