missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) bromide, ultra dry, 99.99% (REO)

Produktkod. 15472907
Change view
Klicka för att se tillgängliga alternativ
Quantity:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
15472907 5 g 5g
15462907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15472907 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 047196.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14456-51-0 | Br3Tm | 408.646 g/mol

Thulium(III) bromide is used for development of advanced ceramic discharge lamps1 and metal halide lamp. Reagent for complexation of lanthanide bromides with aluminum bromide. Reactant for preparation of alkali metal thulium bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14456-51-0
Molecular Formula Br3Tm
Molecular Weight (g/mol) 408.646
MDL Number MFCD00050138
InChI Key HQSWGSFQSCMHFQ-UHFFFAOYSA-K
Synonym thulium bromide, thulium tribromide, thulium iii bromide, thulium 3+ tribromide, acmc-1bo43, thulium iii bromide, anhydrous, ampuled under argon trace metals basis 1g
PubChem CID 84454
IUPAC Name thulium(3+);tribromide
SMILES [Br-].[Br-].[Br-].[Tm+3]

Specifications

Boiling Point 1440°C
Melting Point 952°C
Physical Form Beads
Quantity 5 g
Packaging Plastic bottle; Ampouled under argon
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 408.66
Percent Purity 99.99%
Chemical Name or Material Thulium(III) bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.