missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thulium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

Product Code. 11480281
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480281 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11480281 Supplier Alfa Aesar™ Supplier No. 040565.NA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14014-10-9
Molecular Formula Cl3O12Tm
Molecular Weight (g/mol) 467.272
MDL Number MFCD00049609
InChI Key JNNWMYZIHRZEKX-UHFFFAOYSA-K
Synonym thulium 3+ perchlorate, thulium iii perchlorate, perchloric acid,thulium 3+ salt 8ci,9ci, acmc-1bs9x, perchloric acid thulium iii salt, thulium 3+ triperchlorate, thulium iii perchlorate w/w aqueous solution, reagent grade
PubChem CID 15335706
IUPAC Name thulium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Tm+3]

Specifications

Physical Form Liquid
Quantity 2 g
UN Number UN3211
Formula Weight 467.28
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Density 1.51g/mL
Chemical Name or Material Thulium(III) perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.