missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) sulfate octahydrate, 99.99%, Thermo Scientific™

Product Code. 11413537
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11413537 5 g 5g
11403537 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11413537

Brand: Thermo Scientific Alfa Aesar A17296.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a reactant in dehydration reactions. It is the important dopant for fiber amplifiers.

Solubility
Soluble in water, moderately soluble in strong mineral acids.

Notes
Store at room temperature. Keep away from strong oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13778-40-0
Molecular Formula H16O20S3Tm2
Molecular Weight (g/mol) 770.156
MDL Number MFCD00149881
InChI Key GPBYLGMJMIXKMU-UHFFFAOYSA-H
Synonym thulium sulfate octahydrate, unii-8zgz55l20j, thulium sulfate octahydrate mi, acmc-20aji8, thulium sulfate tm2 so4 3 octahydrate, thulium sulfate hydrate tm2 so4 3.8h2o, sulfuric acid, thulium 3+ salt 3:2 , octahydrate, thulium sulfate-water 2/3/8, thulium sulfate,, sulfuric acid thulium iii salt octahydrate
PubChem CID 71310074
IUPAC Name thulium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Tm+3].[Tm+3]

Specifications

Quantity 5 g
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
Formula Weight 770.17 (626.06 Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Chemical Name or Material Thulium(III) sulfate octahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.