missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymidine 5'-Diphosphate Trisodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Thymidine 5'-Diphosphate Trisodium Salt |
| CAS | 95648-78-5 |
| Molecular Formula | C10 H13 N2 Na3 O11 P2 |
| Quantity | 50 mg |
| Synonym | Thymidine 5'-(Trihydrogen Diphosphate) Trisodium Salt |
| InChI Formula | InChI=1S/C10H16N2O11P2.3Na/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(22-8)4-21-25(19,20)23-24(16,17)18;;;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,11,14,15)(H2,16,17,18);;;/q;3*+1/p-3/t6-,7+,8+;;;/m0.../s1 |
| SMILES | CC1=CN([C@H]2C[C@H](O)[C@@H](COP(=O)(O[Na])OP(=O)(O[Na])O[Na])O2)C(=O)NC1=O |
| IUPAC Name | [disodiooxyphosphoryloxy-[[(2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methoxy]phosphoryl]oxysodium |
| Molecular Weight (g/mol) | 468.134 |
| Formula Weight | 467.969 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?