missing translation for 'onlineSavingsMsg'
Learn More

Thymidine 5'-Diphosphate Trisodium Salt, TRC

Product Code. 30422711
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30422711 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30422711

Brand: Toronto Research Chemicals TRCT077195250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thymidine 5'-Diphosphate Trisodium Salt
CAS 95648-78-5
Molecular Formula C10 H13 N2 Na3 O11 P2
Quantity 250 mg
Synonym Thymidine 5'-(Trihydrogen Diphosphate) Trisodium Salt
InChI Formula InChI=1S/C10H16N2O11P2.3Na/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(22-8)4-21-25(19,20)23-24(16,17)18;;;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,11,14,15)(H2,16,17,18);;;/q;3*+1/p-3/t6-,7+,8+;;;/m0.../s1
SMILES CC1=CN([C@H]2C[C@H](O)[C@@H](COP(=O)(O[Na])OP(=O)(O[Na])O[Na])O2)C(=O)NC1=O
IUPAC Name [disodiooxyphosphoryloxy-[[(2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methoxy]phosphoryl]oxysodium
Molecular Weight (g/mol) 468.134
Formula Weight 467.969
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.